missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Androgen Receptor (pSer216) Rabbit Polyclonal Antibody with BSA and azide
Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Androgen Receptor, Rabbit Polyclonal Antibody.
£117.00 - £586.50
Specifications
Antigen | Androgen Receptor |
---|---|
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12693357
|
Epredia™
RB-9030-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12603367
|
Epredia™
RB-9030-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12683357
|
Epredia™
RB-9030-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12633367
|
Epredia™
RB-9030-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The androgen receptor (AR) is a member of the steroid superfamily of ligand-dependent transcription factors. The expression of AR is reportedly inversely correlated with histologic grade, i.e. well-differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: N-terminal
Immunogen: A synthetic peptide derived from N-terminal of rat AR
Molecular Weight: 110kDa
Positive Control: LnCAp cells or prostate CA
Cellular Localization: Nuclear
This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Specifications
Androgen Receptor | |
Polyclonal | |
Rabbit | |
Oncogenesis | |
A synthetic peptide derived from N-terminal of rat AR | |
Primary |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
IVD | |
Human, Mouse | |
IgG |