Learn More
Anandamide, Tocris Bioscience™
Endogenous and non-selective CB agonist
Brand:  Tocris (Bio-Techne) 13395
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 5mg
 View more versions of this product
Product Code. 13429183
  • £97.02 / 5mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Endogenous cannabinoid and TRPV1 receptor agonist (Ki values are 89 and 371 nM at CB1 and CB2 receptors respectively; EC50 values are 18, 31 and 27 nM at GPR55, CB1 and CB2 respectively; pKi = 5.68 for rTRPV1). Also blocks TNF-α-induced NF-κB activation via direct inhibition of IKK.
Available in water soluble emulsion . Deuterated analog also available.
Specifications
Chemical Identifiers
94421-68-8
347.543
anandamide, arachidonylethanolamide, arachidonoyl ethanolamide, anandamide 20.4, n-6, n-arachidonoyl ethanolamine, arachidonoylethanolamide, n-arachidonoyl-2-hydroxyethylamide, arachidonoyl-ea, aea, n-arachidonoylethanolamine
CHEBI:2700
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO
C22H37NO2
LGEQQWMQCRIYKG-DOFZRALJSA-N
5281969
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
Specifications
5mg
94421-68-8
anandamide, arachidonylethanolamide, arachidonoyl ethanolamide, anandamide 20.4, n-6, n-arachidonoyl ethanolamine, arachidonoylethanolamide, n-arachidonoyl-2-hydroxyethylamide, arachidonoyl-ea, aea, n-arachidonoylethanolamine
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO
347.543
CHEBI:2700
>98%
Anandamide
C22H37NO2
LGEQQWMQCRIYKG-DOFZRALJSA-N
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
5281969
347.54