Learn More
Anandamide (in Tocrisolve 100), Tocris Bioscience™
Endogenous CB receptor agonist (in water-soluble emulsion)
Brand:  Tocris (Bio-Techne) 101750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13448303
  • £771.75 / 50mg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
Endogenous cannabinoid and vanilloid receptor agonist, in water-soluble emulsion (for details see TocrisolveTM 100, Cat. No. 1684). Ki values are 89 and 371 nM for CB1 and CB2 receptors respectively; EC50 values are 18, 31 and 27 nM at GPR55, CB1 and CB2 respectively; pKi = 5.68 for rVR1. Also blocks TNF-α-induced NF-kB activation via direct inhibition of IKK.
Available as the pure oil dissolved in ethanol, Anandamine . Tocrisolve Control also available.
Specifications
Chemical Identifiers
94421-68-8
347.543
anandamide, arachidonylethanolamide, arachidonoyl ethanolamide, anandamide 20.4, n-6, n-arachidonoyl ethanolamine, arachidonoylethanolamide, n-arachidonoyl-2-hydroxyethylamide, arachidonoyl-ea, aea, n-arachidonoylethanolamine
CHEBI:2700
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO
C22H37NO2
LGEQQWMQCRIYKG-DOFZRALJSA-N
5281969
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
Specifications
50mg
94421-68-8
C22H37NO2
LGEQQWMQCRIYKG-DOFZRALJSA-N
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
5281969
347.54
Anandamide (in Tocrisolve™ 100)
>98%
anandamide, arachidonylethanolamide, arachidonoyl ethanolamide, anandamide 20.4, n-6, n-arachidonoyl ethanolamine, arachidonoylethanolamide, n-arachidonoyl-2-hydroxyethylamide, arachidonoyl-ea, aea, n-arachidonoylethanolamine
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO
347.543
CHEBI:2700