Learn More
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank
Glass Thin Layer Chromatography (TLC) Developing Tank designed for the development of TLC plates.
Brand:  Analtech 34105-00
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11795348
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum.
  • Tanks accommodate standard TLC plates.
  • The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development.
  • Thinline™ tanks accommodate two plates and require less solvent for development.
  • Latch-Lid™ units hold lids in place with stainless steel latches.
  • Order rack to develop maximum number of plates in a tank.
Specifications
Specifications
Glass
1 Ea.
TLC Tank
100 x 100 mm