Learn More
ANKRD13A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560280
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IQQSLLESSR SQELSGPASN GGISQTNTYD AQYERAIQES LLTSTEGLCP SALSETSRFD NDLQLAMELS AKELEEWELR LQEE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.
ANKRD13A Polyclonal antibody specifically detects ANKRD13A in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ANKRD13A | |
Polyclonal | |
Unconjugated | |
ANKRD13A | |
1100001D10Rik; Ankrd13; ANKRD13A; ankyrin repeat domain 13; ankyrin repeat domain 13A; ankyrin repeat domain-containing protein 13A; AU046136; NY-REN-25; NY-REN-25 antigen; Protein KE03 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
88455 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ07 | |
ANKRD13A | |
Recombinant protein corresponding to Human AN13A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only