Learn More
Amthamine dihydrobromide, Tocris Bioscience™
Highly selective standard H2 agonist
Brand:  Tocris (Bio-Techne) 066810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487863
  • £132.00 / 10mg
Estimated Shipment
28-06-2018
Add to basket
Description
Description
Highly selective H2 agonist, slightly more potent than histamine itself. Only a weak antagonist at H3 and has no activity at H1 receptors. Induces vasodilation of cerebral arteries and decreases myogenic tone in vitro.
Specifications
Chemical Identifiers
Amthamine dihydrobromide
C6H11N3S·2HBr
319.06
2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
Specifications
Amthamine dihydrobromide
>99%
319.06
2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
142457-00-9
C6H11N3S·2HBr