missing translation for 'onlineSavingsMsg'
Learn More

AMTB hydrochloride, Tocris Bioscience™

TRPM8 blocker

Brand:  Tocris (Bio-Techne) 398910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443934

  • £188.76 / 10mg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TRPM8 channel blocker. Inhibits icilin -induced TRPM8 channel activation in a rat model (pIC50 = 6.23).
Specifications

Chemical Identifiers

926023-82-7
430.991
amtb hydrochloride, n-3-aminopropyl-2-3-methylbenzyl oxy-n-thiophen-2-ylmethyl benzamide hydrochloride, n-3-aminopropyl-2-3-methylphenyl methoxy-n-2-thienylmethyl benzamide hydrochloride, n-3-aminopropyl-2-3-methylphenyl methyl oxy-n-2-thienylmethyl benzamide hydrochloride salt
N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-(thiophen-2-ylmethyl)benzamide;hydrochloride
C23H27ClN2O2S
UDXGBANGPYONOK-UHFFFAOYSA-N
16095383
CC1=CC(=CC=C1)COC2=CC=CC=C2C(=O)N(CCCN)CC3=CC=CS3.Cl

Specifications

AMTB hydrochloride
926023-82-7
amtb hydrochloride, n-3-aminopropyl-2-3-methylbenzyl oxy-n-thiophen-2-ylmethyl benzamide hydrochloride, n-3-aminopropyl-2-3-methylphenyl methoxy-n-2-thienylmethyl benzamide hydrochloride, n-3-aminopropyl-2-3-methylphenyl methyl oxy-n-2-thienylmethyl benzamide hydrochloride salt
CC1=CC(=CC=C1)COC2=CC=CC=C2C(=O)N(CCCN)CC3=CC=CS3.Cl
430.991
430.99
10mg
C23H27ClN2O2S
UDXGBANGPYONOK-UHFFFAOYSA-N
N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-(thiophen-2-ylmethyl)benzamide;hydrochloride
16095383
>99%
Videos