Learn More
AMT hydrochloride, Tocris Bioscience™
Potent, selective iNOS inhibitor
Brand:  Tocris (Bio-Techne) 087110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488083
  • £114.00 / 10mg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Potent, selective and reversible inhibitor of iNOS (IC50 = 3.6 nM; approximately 30 and 40 times selective over nNOS and eNOS respectively).
Specifications
Chemical Identifiers
AMT hydrochloride
C5H10N2S·HCl
166.67
2-Amino-5, 6-dihydro-6-methyl-4H-1, 3-thiazine hydrochloride
Specifications
AMT hydrochloride
>98%
C5H10N2S·HCl
2-Amino-5, 6-dihydro-6-methyl-4H-1, 3-thiazine hydrochloride
21463-31-0
10mg
166.67
Videos