missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amnionless Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Amnionless/AMN |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Amnionless Val20-Ala358 Accession # Q9BXJ7 |
Gene ID (Entrez) | 81693 |
Gene Accession No. | Q9BXJ7 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
Amnionless/AMN Polyclonal specifically detects Amnionless/AMN in Human samples. It is validated for Western Blot.Specifications
Amnionless/AMN | |
81693 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Amnionless in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human Amnionless Val20-Ala358 Accession # Q9BXJ7 | |
Q9BXJ7 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only