Learn More
AMN 082 dihydrochloride, Tocris Bioscience™
The first selective mGlu7 agonist
Brand:  Tocris (Bio-Techne) 238510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13461114
  • £114.75 / 10mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
The first selective mGlu7 agonist. Potently inhibits cAMP accumulation and stimulates GTPγS binding in recombinant cells and on membranes expressing mGlu7 (EC50 = 64 - 290 nM). Selective over other mGluR subtypes and selected ionotropic glutamate receptors up to 10 μM. Acts via a novel allosteric site and is orally active and brain penetrant. Reduces haloperidol-induced catalepsy in rats.
Specifications
Chemical Identifiers
97075-46-2
465.462
amn082 dihydrochloride, amn 082 dihydrochloride, n,n'-dibenzhydrylethane-1,2-diamine dihydrochloride, amn hplc , solid, n,n'-bis diphenylmethyl-1,2-ethanediamine dihydrochloride, diphenylmethyl 2-diphenylmethyl amino ethyl amine dihydrochloride, n,n inverted exclamation marka-dibenzhydrylethane-1,2-diamine dihydrochloride
N,N'-dibenzhydrylethane-1,2-diamine;dihydrochloride
C28H30Cl2N2
YRQCDCNQANSUPB-UHFFFAOYSA-N
11698390
C1=CC=C(C=C1)C(C2=CC=CC=C2)NCCNC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
Specifications
AMN 082 dihydrochloride
97075-46-2
amn082 dihydrochloride, amn 082 dihydrochloride, n,n'-dibenzhydrylethane-1,2-diamine dihydrochloride, amn hplc , solid, n,n'-bis diphenylmethyl-1,2-ethanediamine dihydrochloride, diphenylmethyl 2-diphenylmethyl amino ethyl amine dihydrochloride, n,n inverted exclamation marka-dibenzhydrylethane-1,2-diamine dihydrochloride
C1=CC=C(C=C1)C(C2=CC=CC=C2)NCCNC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl
465.462
465.45
10mg
C28H30Cl2N2
YRQCDCNQANSUPB-UHFFFAOYSA-N
N,N'-dibenzhydrylethane-1,2-diamine;dihydrochloride
11698390
>99%
Videos