missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium tungsten oxide pentahydrate, 99.999% (metals basis), Thermo Scientific™, Puratronic™
Click to view available options
Quantity:
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 99.999% (Metal basis) |
Solubility Information | Slightly soluble in water. |
Formula Weight | 3132.64 (3042.56 Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Density | 2.3 |
Chemical Name or Material | Ammonium tungsten oxide pentahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'