missing translation for 'onlineSavingsMsg'
Learn More

Ammonium trifluoromethanesulfonate, 99%, ACROS Organics™

Brand:  Acros Organics 388730500

Code : 29

Additional Details : CAS Number : 38542-94-8 Weight : 0.10000kg

Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10608434

  • £182.80 / 50g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Ammonium trifluoromethanesulfonate
CH4F3NO3S
MFCD00075332
99%
167.11
Ammonium triflate

Specifications

Ammonium trifluoromethanesulfonate
Crystalline Powder
CH4F3NO3S
MFCD00075332
98.5% min.
Glass bottle
50g
38542-94-8
99%
167.11
Ammonium triflate
Authentic
White
Videos
SDS