missing translation for 'onlineSavingsMsg'
Learn More

Ammonium trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog Number p-3756605
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
38542-94-8
CH4F3NO3S
167.11
MFCD00075332
BMWDUGHMODRTLU-UHFFFAOYSA-N
ammonium trifluoromethanesulfonate, ammonium triflate, acmc-20alsc, methanesulfonic acid,1,1,1-trifluoro-, ammonium salt 1:1
15842047
azanium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[NH4+]
This item is not returnable. View return policy
38542-94-8
CH4F3NO3S
167.11
MFCD00075332
BMWDUGHMODRTLU-UHFFFAOYSA-N
ammonium trifluoromethanesulfonate, ammonium triflate, acmc-20alsc, methanesulfonic acid,1,1,1-trifluoro-, ammonium salt 1:1
15842047
azanium;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[NH4+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 225°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Formula Weight 167.11
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Ammonium trifluoromethanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'