missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10111900
47.43 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


79.95 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
250 g
500 g
1 kg
1762-95-4
CH4N2S
76.12
11428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium cyanosulfanide
[NH4+].[S-]C#N
1762-95-4
CH4N2S
76.12
11428
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium cyanosulfanide
[NH4+].[S-]C#N

CH4N2S, CAS Number-1762-95-4, 500g, White, 76.12g/mol, 149 deg.C, 11428, Plastic powder jar, 5 to 6, Solid

TRUSTED_SUSTAINABILITY
Melting Point 149°C
Color White
pH 5 to 6
Physical Form Solid
Quantity 250 g
Formula Weight 76.12g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Ammonium thiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P233-Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'