missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate, Certified AR for Analysis, Fisher Chemical™
47.43 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
CH4N2S, CAS Number-1762-95-4, 500g, White, 76.12g/mol, 149 deg.C, 11428, Plastic powder jar, 5 to 6, Solid
Specifications
Melting Point | 149°C |
Color | White |
pH | 5 to 6 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 76.12g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Ammonium thiocyanate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P233-Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'