missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrathiomolybdate, 99.95%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-4509437
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15060-55-6
H10MoN2S4
262.28
MFCD00136013
ZKKLPDLKUGTPME-UHFFFAOYSA-N
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
15251598
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
This item is not returnable. View return policy
15060-55-6
H10MoN2S4
262.28
MFCD00136013
ZKKLPDLKUGTPME-UHFFFAOYSA-N
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
15251598
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 300.0°C
Color White
Physical Form Crystals or Powder
Quantity 1 g
Assay Percent Range 99.95%
Linear Formula (NH4)2MoS4
Merck Index 15,557
IUPAC Name diazanium;bis(sulfanylidene)molybdenum;sulfanide
Formula Weight 260.28
Percent Purity 99.95%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium tetrathiomolybdate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium tetrathiomolybdate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'