missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ammonium tetrafluoroborate, 99.999% (metals basis)

Brand:  Alfa Aesar™ 044055.14

Code : NEW

Additional Details : CAS Number : 13826-83-0 Weight : 0.02500kg Transport : UN number : 3260 Chem class : 8 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11315128

  • £72.00 / 25g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Ammonium tetrafluoroborate
NH4BF4
MFCD06245785
13826-83-0
104.84
UN3260

Specifications

Ammonium tetrafluoroborate
Crystalline
(metals basis)
104.84
UN3260
Ammonia-like
220°C (subl.)
13826-83-0
99.999%
NH4BF4
MFCD06245785
1.871g/mL
White
25g
Videos
SDS