missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number 11327157
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
13826-83-0
BF4H4N
104.84
MFCD06245785
PDTKOBRZPAIMRD-UHFFFAOYSA-O
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
9964072
[NH4+].F[B-](F)(F)F
This item is not returnable. View return policy
13826-83-0
BF4H4N
104.84
MFCD06245785
PDTKOBRZPAIMRD-UHFFFAOYSA-O
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
9964072
[NH4+].F[B-](F)(F)F

Ammonium fluoroborate is used as an analytical reagent. In the textile printing and dyeing industry, it used as a catalyst for resin finishing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220°C (sublimation)
Color White
Physical Form Powder
Quantity 500 g
UN Number UN3260
Solubility Information Soluble in water.
IUPAC Name azanium;tetrafluoroborate
Formula Weight 104.84
Percent Purity 97%
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.871 g/mL
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'