missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min, Thermo Scientific Chemicals

Catalog Number p-7048624
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13820-41-2
Cl4H8N2Pt
372.96
MFCD00010885
QJIMNDWDOXTTBR-UHFFFAOYSA-L
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
platinum(2+) diammonium tetrachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]
This item is not returnable. View return policy
13820-41-2
Cl4H8N2Pt
372.96
MFCD00010885
QJIMNDWDOXTTBR-UHFFFAOYSA-L
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
platinum(2+) diammonium tetrachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]

Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 145°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 1 g
Linear Formula (NH4)2PtCl4
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 372.98
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 51% min.
Odor Odorless
Density 2.936 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'