missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 98% |
Linear Formula | (NH4)2PdCl4 |
Fieser | 03,81; 01,778; 02,303; 04,379; 05,496; 06,442; 09,344; 10,297; 11,389; 12,367; 13,103; 15,245; 16,259; 17,255 |
Solubility Information | Solubility in water: soluble |
IUPAC Name | palladium(2+) diammonium tetrachloride |
Formula Weight | 284.28 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Ammonium tetrachloropalladate(II)
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'