missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 40-44% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number p-4407236
Click to view available options
Quantity:
100 mL
500 mL
12135-76-1
H8N2S
68.14
MFCD00010892
UXXGWUQNRMPNKH-UHFFFAOYSA-N
diamine sulfane
N.N.S
12135-76-1
H8N2S
68.14
MFCD00010892
UXXGWUQNRMPNKH-UHFFFAOYSA-N
diamine sulfane
N.N.S

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth.It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Orange
Physical Form Liquid
Quantity 100 mL
Solubility Information Miscible in water,alcohol and liquid ammonia.
Formula Weight 68.14
Odor Stench
Appearance Yellow to orange liquid
Vapor Pressure 546 hPa (410mm Hg) at 20°C
Chemical Name or Material Ammonium sulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'