missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Protein Precipitation, Liquid Chromatography and Molecular Biology Applications
Specifications
Quantity | 5 kg |
Formula Weight | 132.14 |
Grade | Ultrapure Bioreagent |
Packaging | Poly Pail |
Specific Gravity | 1L = 1.77kg |
Chemical Name or Material | Ammonium Sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'