missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number 15557864
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

CAS 7783-20-2
Physical Form Granules
Quantity 2.5 kg
Molecular Formula H8N2O4S
MDL Number MFCD00003391
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name sulfuric acid diamine
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
ChEBI CHEBI:62946
Formula Weight 132.14
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.77kg
Chemical Name or Material Ammonium Sulfate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'