missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number 15109293
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
N.N.OS(O)(=O)=O
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
N.N.OS(O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280.0°C
Color Silver to Gray
pH 5.0 to 6.0 (5% soln. at 25°C)
Physical Form Granules, Fillings or Chips
Quantity 25 kg
Assay Percent Range 99+%
Linear Formula (NH4)2SO4
Merck Index 15, 552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
IUPAC Name diazanium;sulfate
Formula Weight 132.14
Percent Purity ≥99%
Grade ACS Reagent
Packaging Fibre drum
Residue after Ignition 0.005 % max.
Chemical Name or Material Ammonium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'