missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, ACS, 98% min, Thermo Scientific Chemicals

Catalog Number 11302488
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]

Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. It is used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 160°C (decomposition)
Melting Point 131°C to 135°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98% min.
Solubility Information Very soluble in water,liquid ammonia. Slightly soluble in ethanol,glycerol,glycol
Formula Weight 114.12
Grade ACS Reagent
Sensitivity Hygroscopic
Chemical Name or Material Ammonium sulfamate, For ACS analysis

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'