missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfamate, ACS, 98% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. It is used as a compost accelerator, flame retardant and in industrial processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 160°C (decomposition) |
Melting Point | 131°C to 135°C |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 98% min. |
Solubility Information | Very soluble in water,liquid ammonia. Slightly soluble in ethanol,glycerol,glycol |
Formula Weight | 114.12 |
Grade | ACS Reagent |
Sensitivity | Hygroscopic |
Chemical Name or Material | Ammonium sulfamate, For ACS analysis |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'