missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sodium phosphate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals
94.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Melting Point | 80°C |
Color | Colorless to White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99% |
Linear Formula | Na(NH4)HPO4·4H2O |
Merck Index | 12,587 |
Formula Weight | 209.07 |
Percent Purity | 99% |
Grade | Analytical |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'