missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphomolybdate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number p-7049674
Click to view available options
Quantity:
100 g
500 g
54723-94-3
H80Mo12N3O41P-6
1960.994
MFCD00036312
BIQVMZMYTFVCJZ-UHFFFAOYSA-H
ammonium phosphomolybdate hydrate
131664298
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]
54723-94-3
H80Mo12N3O41P-6
1960.994
MFCD00036312
BIQVMZMYTFVCJZ-UHFFFAOYSA-H
ammonium phosphomolybdate hydrate
131664298
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]

Ammonium phosphomolybdate hydrate is used to prepare aluminosilicate-supported heteropolyacid catalysts, which finds application in the dehydration of glycerol in the gas phase to get acrolein. It is used as an inorganic ion exchanger involved in the removal of cesium from liquid wastes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Yellow
Physical Form Powder
Quantity 100 g
Linear Formula (NH4)3PO4·12MoO3·xH2O (x≈3)
Solubility Information Slightly soluble in water.
Formula Weight 1876.35 (Anhydrous)
Grade Reagent
Density 3.15 g/mL
Chemical Name or Material Ammonium phosphomolybdate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'