missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
CAS | 7722-76-1 |
Quantity | 500 g |
Molecular Formula | H6NO4P |
MDL Number | MFCD00003396 |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
SMILES | N.OP(O)(O)=O |
IUPAC Name | azanium;dihydrogen phosphate |
Molecular Weight (g/mol) | 115.03 |
PubChem CID | 24402 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'