missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphate, dibasic, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog Number p-4440131
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
N.N.OP(O)(O)=O
This item is not returnable. View return policy
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
N.N.OP(O)(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 g
Assay Percent Range 99%
Linear Formula (NH4)2HPO4
Merck Index 15,54
Solubility Information (1 M in water) Clear colorless
IUPAC Name diazanium;hydrogen phosphate
Formula Weight 132.06
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Ammonium phosphate, dibasic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'