missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium phosphate, dibasic, 99+%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Color | White |
pH | 7.9 to 8.3 (5% soln. at 20°C) |
Physical Form | Adhering Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99+% |
Linear Formula | (NH4)2HPO4 |
Merck Index | 15, 540 |
Solubility Information | Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone |
Formula Weight | 132.06 |
Percent Purity | ≥99% |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'