missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | (NH4)2S2O8 |
UN Number | 1444 |
Solubility Information | Soluble in water |
IUPAC Name | diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
Formula Weight | 228.2g/mol |
Percent Purity | ≥98% |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.05% (as SO4) |
Density | Vapor Density: 7.9 (vs air) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'