missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™

Catalog Number 10081503
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


30.17 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis)

Electrophoresis-grade Ammonium Persulfate is an initiator of acrylamide polymerization for the preparation of polyacrylamide electrophoresis gels.
TRUSTED_SUSTAINABILITY
Melting Point 120°C
Color White
pH 3.2
Physical Form Powder/Solid
Quantity 25 g
Assay Percent Range ≥98.0 %
Linear Formula (NH4)2S2O8
Merck Index 15, 539
Formula Weight 228.19
Percent Purity ≥98.0%
Grade Electrophoresis
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure Negligible
Decomposition Information >120°C
Density 1.980g/cm³
Chemical Name or Material Ammonium Persulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium Persulfate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'