missing translation for 'onlineSavingsMsg'
Learn More

Ammonium peroxydisulfate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog Number p-4407230
Click to view available options
Quantity:
100 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Ammonium peroxydisulfate is used as an oxidizer, a bleaching agent, in the field of photography, to make aniline dyes, electroplating and in analytical chemistry for determination of manganese.Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 120°C (decomposition)
Physical Form Lump
Quantity 100 g
Assay Percent Range 98.0% min
Flash Point None
Solubility Information Freely soluble in water.
Formula Weight 228.19
Grade ACS
Refractive Index 1.5
Sensitivity Moisture sensitive
Density 1.982 g/mL
Chemical Name or Material Ammonium peroxydisulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'