missing translation for 'onlineSavingsMsg'
Learn More

Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals

Catalog Number p-7080820
Click to view available options
Quantity:
500 g
1000 g
5000 g
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
diazanium;sulfonatooxy sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

Solubility
Freely soluble in water.

Notes
Aqueous solution decomposes slowly at room temperature.Moisture sensitive. Incompatible with strong reducing agents, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY
Melting Point 120°C (decomposition)
Quantity 500 g
Linear Formula (NH4)2S2O8
UN Number UN1444
Flash Point None
Merck Index 14,541
Solubility Information Freely soluble in water.
Formula Weight 228.19
Percent Purity 98%
Odor Odorless
Refractive Index 1.5
Sensitivity Moisture sensitive
Density 1.982
Chemical Name or Material Ammonium peroxydisulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'