missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 70.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 99+% |
Linear Formula | (CO2NH4)2·H2O |
Merck Index | 15, 534 |
Solubility Information | Solubility in water: 45g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 142.11 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'