missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

Catalog Number p-7050226
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
This item is not returnable. View return policy
6009-70-7
C2H10N2O5
142.111
MFCD00149694
MSMNVXKYCPHLLN-UHFFFAOYSA-N
ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
516808
diazanium;oxalate;hydrate
C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

In detinning of iron, metal polishing

In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
TRUSTED_SUSTAINABILITY
Melting Point 70°C (decomposition)
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.0-101.0%
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water; Slightly soluble in alcohol.
Formula Weight 142.11 (124.10 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Density 1.5
Chemical Name or Material Ammonium Oxalate Monohydrate, Monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ethanedioic acid, diammonium salt, monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'