missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Nitrate, Reagent ACS, Crystals, 98%, ACROS Organics™

Brand:  Acros Organics 423350010

Code : 29

Additional Details : CAS Number : 6484-52-2 Weight : 1.05000kg Transport : UN number : 1942 Chem class : 1 Pack group : III

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10547942

  • £45.70 / 1kg


EU Stock 42
Add to basket

Specifications

Chemical Identifiers

Ammonium nitrate
ACS Reagent Grade
98%
NH4NO3
MFCD00011425
6484-52-2
98%
H4N2O3
80.04

Specifications

Ammonium nitrate
ACS Reagent Grade
98%
H4N2O3
80.04
01,39; 15,14
5ppm max.
5ppm max.
2ppm max.
Plastic bottle
0.01% max.
1.72
210.0°C
169.0°C
1kg
6484-52-2
Adhering Crystals
98%
NH4NO3
MFCD00011425
15, 531
1.7200g/mL
0.005% max.
Passes Test (limit about 5ppm)
5ppm max.
Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol
20ppm max.
White
4.5 to 6.0 (5% soln. at 25°C)
Videos
SDS