missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate, 99+%, for analysis, ACROS Organics™

Brand:  Acros Organics 205865000

Code : 29

Additional Details : CAS Number : 6484-52-2 Weight : 0.58000kg Transport : UN number : 1942 Chem class : 1 Pack group : III

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10412211

  • £34.90 / 500g


EU Stock 46
Add to basket

Description

Description

For analysis
Specifications

Chemical Identifiers

Ammonium nitrate
6484-52-2
≥99%
H4N2O3
80.04
p.a.
For analysis
99+%
NH4NO3
MFCD00011425

Specifications

Ammonium nitrate
6484-52-2
Adhering Crystals
99+%
NH4NO3
MFCD00011425
1.7200g/mL
2ppm max.
Plastic bottle
Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol
20ppm max.
White
4.5 to 6 (5% soln. at 20°C)
p.a.
For analysis
≥99%
H4N2O3
80.04
5ppm max.
5ppm max.
5ppm max.
0.01% max. (as SO4)
1.72
210.0°C
169.0°C
500g
Videos
SDS