missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Brand:  Alfa Aesar 010638.30

Code : NEW

Additional Details : CAS Number : 6484-52-2 Weight : 0.25000kg Transport : UN number : 1942 Chem class : 1 Pack group : III

Quantity 250g

 View more versions of this product

Product Code. 11360829

  • £286.70 / 250g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Ammonium nitrate
Crystalline
H4N2O3
MFCD00011425
6484-52-2
100.00%
80.04

Specifications

Ammonium nitrate
Crystalline
H4N2O3
80.04
Odorless
250g
6484-52-2
100.00%
Colorless crystalline
MFCD00011425
Colorless
Videos
SDS