Learn More
Ammonium nitrate, 99.999%, (trace metal basis), Acros Organics™
Brand: Acros Organics 436961000
Code : 29
Additional Details : CAS Number : 6484-52-2 Weight : 0.15000kg Transport : UN number : 1942 Chem class : 1 Pack group : III
Quantity | 100g |
---|---|
Packaging | Plastic bottle |
Description
For ultra trace analysisChemical Identifiers
Ammonium nitrate | |
Trace Metal Basis | |
H4N2O3 | |
80.04 |
6484-52-2 | |
99.999% | |
NH4NO3 |
Specifications
Ammonium nitrate | |
Trace Metal Basis | |
99.999% | |
H4N2O3 | |
80.04 | |
Plastic bottle | |
1.72 | |
210.0°C | |
169.0°C |
6484-52-2 | |
Crystalline Powder | |
99.999% | |
NH4NO3 | |
1.7200g/mL | |
Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol | |
10ppm max. | |
White | |
100g |