missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate, 99.999%, (trace metal basis), ACROS Organics™

Brand:  Acros Organics 436960250

Code : 29

Additional Details : CAS Number : 6484-52-2 Weight : 0.08000kg Transport : UN number : 1942 Chem class : 5.1 Pack group : III

Packaging Plastic bottle
Quantity 25g

 View more versions of this product

Product Code. 10058339

  • £57.77 / 25g

In stock, ready to ship: 9

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For ultra trace analysis
Specifications

Chemical Identifiers

6484-52-2
80.04
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
CHEBI:63038
[NH4+].[N+](=O)([O-])[O-]
H4N2O3
DVARTQFDIMZBAA-UHFFFAOYSA-O
22985
azanium;nitrate

Specifications

Ammonium nitrate
99.999%
NH4NO3
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
DVARTQFDIMZBAA-UHFFFAOYSA-O
azanium;nitrate
22985
80.04
99.999%
1.7200g/mL
Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol
10ppm max.
White
25g
6484-52-2
H4N2O3
15,531
Solubility in water: 190g/100mL (20°C). Other solubilities: 1g/20mL alcohol,1g/8mL methanol
[NH4+].[N+](=O)([O-])[O-]
80.04
CHEBI:63038
Crystalline Powder
Trace Metal Basis
Plastic bottle
1.72
210.0°C
169.0°C
Videos
SDS Documents