missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4509574
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
12054-85-2
H24Mo7N6O24·4H2O
1235.86
12054-85-2
H24Mo7N6O24·4H2O
1235.86

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY
Melting Point 190.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min
Linear Formula (NH4)6Mo7O24·4H2O
Merck Index 15,53
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 1235.86
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Ammonium molybdate(VI) tetrahydrate
Show More Show Less
Product Identifier
  • Ammonium molybdate(VI) tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'