missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, Honeywell

Catalog Number p-7072207
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
12054-85-2
MFCD00167059
This item is not returnable. View return policy
12054-85-2
MFCD00167059
Quantity 50 g
Linear Formula (NH4)6Mo7O24 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 1235.86g/mol
Concentration or Composition (by Analyte or Components) 81 to 83% MoO3 basis, 99.98% (trace metals basis)
Density 2.498g/mL at 25°C
Chemical Name or Material Ammonium molybdate tetrahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'