missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
53.41 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
90.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21741" to get your promotional price.
90.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Description
H24MO7N6O24.4H2O, CAS Number-12054-85-2, 500g, White, 1235.86g/mol, 190 deg.C, 167059, Plastic powder jar, 5.0 to 5.5, Solid
Specifications
Melting Point | 190°C |
Color | White |
pH | 5.0 to 5.5 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 1235.86g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Ammonium molybdate(VI) tetrahydrate
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'