missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate tetrahydrate, ACS Reagent, Honeywell Fluka™

Brand:  Honeywell Fluka A7302-1KG

Code : NEW

Additional Details : CAS Number : 12054-85-2 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15637670

  • £193.93 / 1kg


Estimated Shipment date
from Supplier 26-01-2018
Add to basket

Specifications

Chemical Identifiers

12054-85-2
81 to 83% (MoO3 basis)
1235.86g/mol
(NH4)6Mo7O24 · 4H2O
NONH for all modes of transport
Ammonium molybdate tetrahydrate
2.498g/mL at 25°C
ACS Reagent
MFCD00167059

Specifications

Ammonium molybdate tetrahydrate
ACS Reagent
1235.86g/mol
NONH for all modes of transport
2.498g/mL at 25°C
12054-85-2
(NH4)6Mo7O24 · 4H2O
MFCD00167059
81 to 83% (MoO3 basis)
1kg
Videos
SDS