missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron (II) Sulfate Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™

£73.03 - £123.00

Chemical Identifiers

CAS 7783-85-9
MDL Number 51039
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10060180
SDS View Documents
Fisher Chemical
J/1101/15
1L Amber glass bottle
£73.03
1 L
View alternative products
Save Up to 
»
In stock, ready to ship: 8
Please to purchase this item. Need a web account? Register with us today!
10375732
SDS View Documents
Fisher Chemical
J/1101/PB17
2.5L HDPE plastic bottle
£123.00
2.50 L
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7783-85-9
51039

Specifications

7783-85-9
Liquid
1L
51039
Amber glass bottle
SDS