Learn More
Ammonium iron(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Ammonium iron(II) sulfate hydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼100°C (decomposition) |
Color | Blue-Green |
Physical Form | Coarse Powder |
Quantity | 5 g |
Linear Formula | (NH4)2Fe(SO4)2·xH2O (x≈6) |
Merck Index | 14,521 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 284.04 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only