missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(II) Sulfate Hexahydrate, Certified AR for analysis, Fisher Chemical™

Catalog Number 10111850
134.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
7783-85-9
FeH20N2O14S2
392.13
MFCD00150530
MQLVWQSVRZVNIP-UHFFFAOYSA-L
iron(3+) diammonium hexahydrate disulfate
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
7783-85-9
FeH20N2O14S2
392.13
MFCD00150530
MQLVWQSVRZVNIP-UHFFFAOYSA-L
iron(3+) diammonium hexahydrate disulfate
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Ammonium Iron(II) Sulfate Hexahydrate, Certified AR for analysis

TRUSTED_SUSTAINABILITY
Melting Point 100°C
Color Green
pH 3,5
Physical Form Solid
Quantity 1 kg
Formula Weight 392.14g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Ammonium iron (II) sulfate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'