missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
122.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Melting Point | 100.0°C |
Color | Green |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 5 kg |
Assay Percent Range | 99+% |
Linear Formula | (NH4)2Fe(SO4)2·6H2O |
Merck Index | 15, 518 |
Solubility Information | Solubility in water: 269g/L (20°C). Other solubilities: practically insoluble in alcohol |
Formula Weight | 392.14 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
Product Identifier
- Ammonium iron (II) sulfate hexahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'